missing translation for 'onlineSavingsMsg'
Learn More

Permethrin, MedChemExpress

Product Code. 16438486
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16438486 100 mg 100mg
16412226 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16438486 Supplier Medchem Express Supplier No. HYB0887
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Permethrin (NRDC-143) is an insecticide, acaricide, and insect repellent; functions as a neurotoxin, affecting neuron membranes by prolonging sodium channel activation.

Permethrin (NRDC-143) is an insecticide, acaricide, and insect repellent

Specifications

Chemical Name or Material Permethrin
CAS 52645-53-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H20Cl2O3
Quantity 100 mg
Synonym NRDC-143
Solubility Information DMSO : 50 mg/mL (127.78 mM; Need ultrasonic)
SMILES O=C(C1C(C)(C)C1/C=C(Cl)\Cl)OCC2=CC=CC(OC3=CC=CC=C3)=C2
Molecular Weight (g/mol) 391.29
Formula Weight 391.29
Percent Purity 98.0%
Grade Research
Physical Form Liquid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.