missing translation for 'onlineSavingsMsg'
Learn More

Perphenazine, MedChemExpress

Product Code. 16452009
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16452009 1 g 1g
16430728 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16452009 Supplier Medchem Express Supplier No. HYA0077
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Perphenazine is an orally active dopamine receptor and histamine-1 receptor antagonist, with Ki values of 0.56 nM (D2), 0.43 nM (D3), 6 nM (5-HT2A), respectively. Perphenazine also binds to Alpha-1A adrenergic receptor. Perphenazine inhibits cancer cell proliferation, and induces apoptosis. Perphenazine can be used in the research of mental disease, cancer, inflammation.

Perphenazine is an orally active dopamine receptor and histamine-1 receptor antagonist, with Ki values of 0.56 nM (D2), 0.43 nM (D3), 6 nM (5-HT2A), respectively. Perphenazine also binds to Alpha-1A adrenergic receptor. Perphenazine inhibits cancer cell proliferation, and induces apoptosis. Perphenazine can be used in the research of mental disease, cancer, inflammation.

Specifications

Chemical Name or Material Perphenazine
Physical Form Powder
CAS 58-39-9
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C21H26ClN3OS
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 100 mg/mL (247.54 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES OCCN1CCN(CCCN2C3=C(C=CC=C3)SC4=CC=C(Cl)C=C24)CC1
Molecular Weight (g/mol) 403.97
Formula Weight 403.97
Percent Purity 99.66%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.