missing translation for 'onlineSavingsMsg'
Learn More

Perrhenic(VII)acid, 99.99%, (trace metal basis), 76.5% solution in water

Product Code. 10419970 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
Product Code. Quantity Packaging unitSize
10419970 5 g Glass bottle 5g
10098821 25 g Glass bottle 25g
2 options
This item is not returnable. View return policy

Product Code. 10419970

Brand: Thermo Scientific Acros 316180050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13768-11-1 | HO4Re | 251.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis, Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13768-11-1
Molecular Formula HO4Re
Molecular Weight (g/mol) 251.21
MDL Number MFCD00011326
InChI Key UGSFIVDHFJJCBJ-UHFFFAOYSA-M
Synonym perrhenic acid, hydroxy trioxo rhenium, perrhenic acid hreo4, rheniumoylol, hydrogen tetraoxorhenate vii, perrhenic vii acid, perrhenic acid solution, acmc-1bqjs, rhenate reo41-, hydrogen, t-4, rhenate reo41-, hydrogen 1:1 , t-4
PubChem CID 83718
IUPAC Name hydroxy(trioxo)rhenium
SMILES O[Re](=O)(=O)=O

Specifications

Color Colorless to Yellow
Assay Percent Range 99.99%
Linear Formula HReO4
Concentration 70 to 76.5% (HReO4)
Density 2.1600g/mL
Formula Weight 251.21
Grade Trace Metal Basis
Packaging Glass bottle
Quantity 5 g
Physical Form Solution
Chemical Name or Material Perrhenic(VII)acid, 99.99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.