missing translation for 'onlineSavingsMsg'
Learn More

Perylene, 99+%

Product Code. p-3758646 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10357060 1 g Glass bottle 1g
10133180 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10357060 Supplier Thermo Scientific Acros Supplier No. 130070010
Add to basket
This item is not returnable. View return policy

CAS: 198-55-0 | C20H12 | 252.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 198-55-0
Molecular Formula C20H12
Molecular Weight (g/mol) 252.31
MDL Number MFCD00004142
InChI Key CSHWQDPOILHKBI-UHFFFAOYSA-N
Synonym peri-dinaphthalene, perilene, dibenz de,kl anthracene, alpha-perylene, perylen, unii-5qd5427un7, ccris 1231, perylen-1-yl, perylen-2-yl
PubChem CID 9142
ChEBI CHEBI:29861
IUPAC Name perylene
SMILES C1=CC2=C3C(=C1)C4=CC=CC5=C4C(=CC=C5)C3=CC=C2

Specifications

Melting Point 277.0°C to 280.0°C
Color Ochre to Brown
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Beilstein 05, I, 363
Merck Index 15, 7299
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in ether, alcohol,, soluble in benzene, toluene,, very soluble in cs2, chloroform, acetone, very sparingly soluble in petroleum ether
Formula Weight 252.31
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material Perylene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.