missing translation for 'onlineSavingsMsg'
Learn More

Sartorius PES Tangential Flow Filtration Cassette for up to 0.5 L

Product Code. 30178716 Shop All Sartorius Products
Click to view available options
MWCO:
10 kDa
100 kDa
1000 kDa
30 kDa
300 kDa
5 kDa
50 kDa
Unit Size:
Pack of 2
This item is not returnable. View return policy

Product Code. 30178716

Brand: Sartorius VFS050P0300IV

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Plug-and-play tangential flow filtration cassette that is made from PES, for ultrafiltration and diafiltration of intermediate sample volumes.

  • Link the cassettes quickly and securely to a suitable pump and reservoirs, right out of the box.
  • All-in-one design avoids the need for expensive cassette holders and TFF systems.
  • Advanced flow path and fixed occlusion restrictor eliminate the effort that is usually needed for TFF.
  • High performance PES or Hydrosart™ RC membranes ensure maximum recoveries.
  • Improve safety for you and your sample with no need for flushing or cleaning.
  • Cassettes contain 30 to 60% less plastic and are supplied with only the tubing you need.
  • Membrane material: PES
  • Recommended feed volume: 100 to 500 mL
TRUSTED_SUSTAINABILITY

Specifications

Filter Type Ultrafiltration
Format Cassette
For Use With (Equipment) Peristaltic Pump
Material Polycarbonate
Material (Membrane) PES
MWCO 300 kDa
Sample Volume 0.1 to 0.5 L
Sterility Non-sterile
Type TFF cassette
Volume (Metric) 0.5 L
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.