missing translation for 'onlineSavingsMsg'
Learn More

Pesticide Mix, 31-1123 100 μg/mL in Acetone, Dr. Ehrenstorfer

Product Code. 30318259
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30318259 5 x 1 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30318259 Supplier Dr. Ehrenstorfer Supplier No. DREGS09000253AC
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (±)-Fenpropathrin∣Alachlor∣alpha-Endosulfan∣Atrazine∣Atrazine-desethyl∣Atrazine-desisopropyl∣Azinphos-methyl∣Benfluralin∣beta-Endosulfan∣Chlorfenapyr∣Chlorothalonil∣Coumaphos∣Cyfluthrin∣Cypermethrin∣Deltamethrin∣Dimethoate∣Fenvalerate∣Fluazifop-P-butyl∣gamma-HCH∣Iprodione∣Malathion∣Metribuzin∣MGK 264∣Parathion-ethyl∣Parathion-methyl∣Permethrin∣Phenothrin∣Phorate∣Prodiamine∣Propyzamide∣Simazine
CAS 39515-41-8, 15972-60-8, 959-98-8, 1912-24-9, 6190-65-4, 1007-28-9
Analyte or Component Names (±)-Fenpropathrin∣Alachlor∣alpha-Endosulfan∣Atrazine∣Atrazine-desethyl∣Atrazine-desisopropyl∣Azinphos-methyl∣Benfluralin∣beta-Endosulfan∣Chlorfenapyr∣Chlorothalonil∣Coumaphos∣Cyfluthrin∣Cypermethrin∣Deltamethrin∣Dimethoate∣Fenvalerate∣Fluazifop-P-butyl∣gamma-HCH∣Iprodione∣Malathion∣Metribuzin∣MGK 264∣Parathion-ethyl∣Parathion-methyl∣Permethrin∣Phenothrin∣Phorate∣Prodiamine∣Propyzamide∣Simazine
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Acetone
Quantity 5 x 1 mL
Concentration or Composition (by Analyte or Components) (±)-Fenpropathrin 100 μg/mL∣Alachlor 100 μg/mL∣alpha-Endosulfan 100 μg/mL∣Atrazine 100 μg/mL∣Atrazine-desethyl 100 μg/mL∣Atrazine-desisopropyl 100 μg/mL∣Azinphos-methyl 100 μg/mL∣Benfluralin 100 μg/mL∣beta-Endosulfan 100 μg/mL∣Chlorfenapyr 100 μg/mL∣Chlorothalonil 100 μg/mL∣Coumaphos 100 μg/mL∣Cyfluthrin 100 μg/mL∣Cypermethrin 100 μg/mL∣Deltamethrin 100 μg/mL∣Dimethoate 100 μg/mL∣Fenvalerate 100 μg/mL∣Fluazifop-P-butyl 100 μg/mL∣gamma-HCH 100 μg/mL∣Iprodione 100 μg/mL∣Malathion 100 μg/mL∣Metribuzin 100 μg/mL∣MGK 264 100 μg/mL∣Parathion-ethyl 100 μg/mL∣Parathion-methyl 100 μg/mL∣Permethrin 100 μg/mL∣Phenothrin 100 μg/mL∣Phorate 100 μg/mL∣Prodiamine 100 μg/mL∣Propyzamide 100 μg/mL∣Simazine 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.