missing translation for 'onlineSavingsMsg'
Learn More

Pesticide Mix C in MeCl2:Acetone:MeOH:MtBE 5:2:2:1, Dr. Ehrenstorfer

Product Code. 30321983
Change view
Click to view available options
Quantity:
4 x 1 mL
Unit Size:
4mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30321983 4 x 1 mL 4mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30321983 Supplier Dr. Ehrenstorfer Supplier No. DREGS09000053DI
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4-Dichlorophenol∣2,4-Dichlorophenoxyacetic acid∣4-(2,4-Dichlorophenoxy)butyric acid∣4-Chloro-2-methylphenol∣Bromoxynil∣Clopyralid∣Dicamba∣Dichlorprop∣Diclofop methyl∣Fluroxypyr∣Hexaconazole∣Imazamethabenz-methyl∣Imazamox∣Imazethapyr∣MCPA∣MCPB∣Mecoprop∣Picloram∣Quinchlorac∣Triclopyr∣Trifluralin
CAS 120-83-2, 94-75-7, 94-82-6, 1570-64-5, 1689-84-5, 1702-17-6
Analyte or Component Names 2,4-Dichlorophenol∣2,4-Dichlorophenoxyacetic acid∣4-(2,4-Dichlorophenoxy)butyric acid∣4-Chloro-2-methylphenol∣Bromoxynil∣Clopyralid∣Dicamba∣Dichlorprop∣Diclofop methyl∣Fluroxypyr∣Hexaconazole∣Imazamethabenz-methyl∣Imazamox∣Imazethapyr∣MCPA∣MCPB∣Mecoprop∣Picloram∣Quinchlorac∣Triclopyr∣Trifluralin
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Dichloromethane
Quantity 4 x 1 mL
Concentration or Composition (by Analyte or Components) 2,4-Dichlorophenol 200 μg/mL∣2,4-Dichlorophenoxyacetic acid 200 μg/mL∣4-(2,4-Dichlorophenoxy)butyric acid 200 μg/mL∣4-Chloro-2-methylphenol 200 μg/mL∣Bromoxynil 200 μg/mL∣Dicamba 200 μg/mL∣Dichlorprop 200 μg/mL∣Diclofop methyl 200 μg/mL∣Hexaconazole 200 μg/mL∣Imazamethabenz-methyl 200 μg/mL∣Imazethapyr 200 μg/mL∣MCPB 200 μg/mL∣Picloram 200 μg/mL∣Quinchlorac 200 μg/mL∣Triclopyr 200 μg/mL∣Trifluralin 200 μg/mL∣Clopyralid 1000 μg/mL∣Fluroxypyr 1000 μg/mL∣Imazamox 1000 μg/mL∣MCPA 1000 μg/mL∣Mecoprop 1000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.