missing translation for 'onlineSavingsMsg'
Learn More

Pesticide Mixture 204 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30312388
Click to view available options
Quantity:
10 x 1 mL
Unit Size:
10mL
This item is not returnable. View return policy

Product Code. 30312388

Brand: Dr. Ehrenstorfer DREGH09000204AL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acetamiprid∣Azoxystrobin∣Bifenazate∣Bifenthrin∣Boscalid∣Buprofezin∣Carbaryl∣Carbofuran∣Chlorantraniliprole∣Chlorfluazuron∣Clothianidin∣Cyprodinil∣Dicrotophos∣Difenoconazole∣Diflubenzuron∣Dimethoate∣Dinotefuran∣Fenbuconazole∣Fenbutatin oxide∣Fipronil∣Flonicamid∣Flubendiamide∣Fludioxonil∣Flufenoxuron∣Hexaflumuron∣Imazalil
CAS 135410-20-7, 131860-33-8, 149877-41-8, 82657-04-3, 188425-85-6, 69327-76-0
Analyte or Component Names Acetamiprid∣Azoxystrobin∣Bifenazate∣Bifenthrin∣Boscalid∣Buprofezin∣Carbaryl∣Carbofuran∣Chlorantraniliprole∣Chlorfluazuron∣Clothianidin∣Cyprodinil∣Dicrotophos∣Difenoconazole∣Diflubenzuron∣Dimethoate∣Dinotefuran∣Fenbuconazole∣Fenbutatin oxide∣Fipronil∣Flonicamid∣Flubendiamide∣Fludioxonil∣Flufenoxuron∣Hexaflumuron∣Imazalil
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile
Quantity 10 x 1 mL
Concentration or Composition (by Analyte or Components) Acetamiprid 100 μg/mL∣Azoxystrobin 100 μg/mL∣Bifenazate 100 μg/mL∣Bifenthrin 100 μg/mL∣Boscalid 100 μg/mL∣Buprofezin 100 μg/mL∣Carbaryl 100 μg/mL∣Carbofuran 100 μg/mL∣Chlorantraniliprole 100 μg/mL∣Chlorfluazuron 100 μg/mL∣Clothianidin 100 μg/mL∣Cyprodinil 100 μg/mL∣Dicrotophos 100 μg/mL∣Difenoconazole 100 μg/mL∣Diflubenzuron 100 μg/mL∣Dimethoate 100 μg/mL∣Dinotefuran 100 μg/mL∣Fenbuconazole 100 μg/mL∣Fenbutatin oxide 100 μg/mL∣Fipronil 100 μg/mL∣Flonicamid 100 μg/mL∣Flubendiamide 100 μg/mL∣Fludioxonil 100 μg/mL∣Flufenoxuron 100 μg/mL∣Hexaflumuron 100 μg/mL∣Imazalil 100 μg/mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.