missing translation for 'onlineSavingsMsg'
Learn More

Pesticide Mixture 205 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30326509
Change view
Click to view available options
Quantity:
10 x 1 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30326509 10 x 1 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30326509 Supplier Dr. Ehrenstorfer Supplier No. DREGH09000205AL

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Imidacloprid∣Lufenuron∣Malathion∣Metalaxyl∣Methidathion∣Methomyl∣Methoxyfenozide∣Novaluron∣Omethoate∣Oxamyl∣Phenthoate∣Prochloraz∣Profenofos∣Propargite∣Pyraclostrobin∣Pyrimethanil∣Spinetoram J∣Spinosad∣Tebuconazole∣Tebufenozide∣Thiabendazole∣Thiacloprid∣Tolfenpyrad∣Triazophos∣Trifloxystrobin
CAS 138261-41-3, 103055-07-8, 121-75-5, 57837-19-1, 950-37-8, 16752-77-5
Analyte or Component Names Imidacloprid∣Lufenuron∣Malathion∣Metalaxyl∣Methidathion∣Methomyl∣Methoxyfenozide∣Novaluron∣Omethoate∣Oxamyl∣Phenthoate∣Prochloraz∣Profenofos∣Propargite∣Pyraclostrobin∣Pyrimethanil∣Spinetoram J∣Spinosad∣Tebuconazole∣Tebufenozide∣Thiabendazole∣Thiacloprid∣Tolfenpyrad∣Triazophos∣Trifloxystrobin
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile
Quantity 10 x 1 mL
Concentration or Composition (by Analyte or Components) Imidacloprid 100 μg/mL∣Lufenuron 100 μg/mL∣Malathion 100 μg/mL∣Metalaxyl 100 μg/mL∣Methidathion 100 μg/mL∣Methomyl 100 μg/mL∣Methoxyfenozide 100 μg/mL∣Novaluron 100 μg/mL∣Omethoate 100 μg/mL∣Oxamyl 100 μg/mL∣Phenthoate 100 μg/mL∣Prochloraz 100 μg/mL∣Profenofos 100 μg/mL∣Propargite 100 μg/mL∣Pyraclostrobin 100 μg/mL∣Pyrimethanil 100 μg/mL∣Spinetoram J 100 μg/mL∣Spinosad 100 μg/mL∣Tebuconazole 100 μg/mL∣Tebufenozide 100 μg/mL∣Thiabendazole 100 μg/mL∣Thiacloprid 100 μg/mL∣Tolfenpyrad 100 μg/mL∣Triazophos 100 μg/mL∣Trifloxystrobin 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.