missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pesticide Mixture 251 100 μg/mL in Acetone, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Allethrin∣Buprofezin∣Carfentrazone-ethyl∣Cyprodinil∣Ethion∣Fenamiphos∣Fipronil∣Fludioxonil∣Flumioxazin∣Flutolanil∣Fosthiazate∣Hexazinone∣Methidathion∣Myclobutanil∣Oxadiazon∣Pendimethalin∣Piperonyl butoxide∣Propiconazole∣Triadimefon∣Tribufos |
| CAS | 584-79-2, 69327-76-0, 128639-02-1, 121552-61-2, 563-12-2, 22224-92-6 |
| Analyte or Component Names | Allethrin∣Buprofezin∣Carfentrazone-ethyl∣Cyprodinil∣Ethion∣Fenamiphos∣Fipronil∣Fludioxonil∣Flumioxazin∣Flutolanil∣Fosthiazate∣Hexazinone∣Methidathion∣Myclobutanil∣Oxadiazon∣Pendimethalin∣Piperonyl butoxide∣Propiconazole∣Triadimefon∣Tribufos |
| Shipping Condition | +20°C |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetone |
| Quantity | 5 x 1 mL |
| Concentration or Composition (by Analyte or Components) | Allethrin 100 μg/mL∣Buprofezin 100 μg/mL∣Carfentrazone-ethyl 100 μg/mL∣Cyprodinil 100 μg/mL∣Ethion 100 μg/mL∣Fenamiphos 100 μg/mL∣Fipronil 100 μg/mL∣Fludioxonil 100 μg/mL∣Flumioxazin 100 μg/mL∣Flutolanil 100 μg/mL∣Fosthiazate 100 μg/mL∣Hexazinone 100 μg/mL∣Methidathion 100 μg/mL∣Myclobutanil 100 μg/mL∣Oxadiazon 100 μg/mL∣Pendimethalin 100 μg/mL∣Piperonyl butoxide 100 μg/mL∣Propiconazole 100 μg/mL∣Triadimefon 100 μg/mL∣Tribufos 100 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?