missing translation for 'onlineSavingsMsg'
Learn More

Pesticide Mixture 254 200 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30326118
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30326118 5 x 1 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30326118 Supplier Dr. Ehrenstorfer Supplier No. DREGS09000254AC
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4-Dichlorophenoxyacetic acid∣Bentazone∣Chloramben∣Clopyralid∣Dicamba∣Imazamox∣Imazapyr∣Imazaquin∣Imazethapyr∣MCPA∣Mecoprop∣Picloram∣Triclopyr
CAS 94-75-7, 25057-89-0, 133-90-4, 1702-17-6, 1918-00-9, 114311-32-9
Analyte or Component Names 2,4-Dichlorophenoxyacetic acid∣Bentazone∣Chloramben∣Clopyralid∣Dicamba∣Imazamox∣Imazapyr∣Imazaquin∣Imazethapyr∣MCPA∣Mecoprop∣Picloram∣Triclopyr
Shipping Condition Ice pack (-20°C)
Certifications/Compliance ISO 17034
Solvent or Matrix Acetone
Quantity 5 x 1 mL
Concentration or Composition (by Analyte or Components) 2,4-Dichlorophenoxyacetic acid 200 μg/mL∣Bentazone 200 μg/mL∣Chloramben 200 μg/mL∣Clopyralid 200 μg/mL∣Dicamba 200 μg/mL∣Imazamox 200 μg/mL∣Imazapyr 200 μg/mL∣Imazaquin 200 μg/mL∣Imazethapyr 200 μg/mL∣MCPA 200 μg/mL∣Mecoprop 200 μg/mL∣Picloram 200 μg/mL∣Triclopyr 200 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.