missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pesticide Mixture 301 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Abamectin∣Acetamiprid∣Amitraz∣Azoxystrobin∣Carbofuran-3-hydroxy∣Chlorantraniliprole∣Fenamidone∣Imazalil∣Imidacloprid∣Methoxyfenozide∣Myclobutanil∣Piperonyl butoxide∣Pyraclostrobin∣Thiabendazole∣Thiophanate-methyl |
| CAS | 71751-41-2, 135410-20-7, 33089-61-1, 131860-33-8, 16655-82-6, 500008-45-7 |
| Analyte or Component Names | Abamectin∣Acetamiprid∣Amitraz∣Azoxystrobin∣Carbofuran-3-hydroxy∣Chlorantraniliprole∣Fenamidone∣Imazalil∣Imidacloprid∣Methoxyfenozide∣Myclobutanil∣Piperonyl butoxide∣Pyraclostrobin∣Thiabendazole∣Thiophanate-methyl |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetonitrile |
| Quantity | 10 mL |
| Concentration or Composition (by Analyte or Components) | Abamectin 100 μg/mL∣Acetamiprid 100 μg/mL∣Amitraz 100 μg/mL∣Azoxystrobin 100 μg/mL∣Carbofuran-3-hydroxy 100 μg/mL∣Chlorantraniliprole 100 μg/mL∣Fenamidone 100 μg/mL∣Imazalil 100 μg/mL∣Imidacloprid 100 μg/mL∣Methoxyfenozide 100 μg/mL∣Myclobutanil 100 μg/mL∣Piperonyl butoxide 100 μg/mL∣Pyraclostrobin 100 μg/mL∣Thiabendazole 100 μg/mL∣Thiophanate-methyl 100 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?