missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pesticide Mixture D in Dichloromethane, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Aminopyralid∣Bentazone∣Clodinafop free acid∣Clodinafop-propargyl ester∣Fluazifop (free acid)∣Iprodione∣Propiconazole∣Quizalofop free acid |
| CAS | 150114-71-9, 25057-89-0, 114420-56-3, 105512-06-9, 69335-91-7, 36734-19-7 |
| Analyte or Component Names | Aminopyralid∣Bentazone∣Clodinafop free acid∣Clodinafop-propargyl ester∣Fluazifop (free acid)∣Iprodione∣Propiconazole∣Quizalofop free acid |
| Shipping Condition | +20°C |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dichloromethane |
| Quantity | 4 x 1 mL |
| Concentration or Composition (by Analyte or Components) | Bentazone 200 μg/mL∣Fluazifop (free acid) 200 μg/mL∣Aminopyralid 1000 μg/mL∣Clodinafop free acid 1000 μg/mL∣Clodinafop-propargyl ester 1000 μg/mL∣Iprodione 1000 μg/mL∣Propiconazole 1000 μg/mL∣Quizalofop free acid 1000 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?