missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pesticide/PCB Surrogate Mixture 55 200 μg/mL in Acetone, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | 2,4,5,6-Tetrachloro-m-xylene∣PCB No. 209 |
| CAS | 877-09-8, 2051-24-3 |
| Analyte or Component Names | 2,4,5,6-Tetrachloro-m-xylene∣PCB No. 209 |
| Shipping Condition | +20°C |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetone |
| Quantity | 10 x 1 mL |
| Concentration or Composition (by Analyte or Components) | 2,4,5,6-Tetrachloro-m-xylene 200 μg/mL∣PCB No. 209 200 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?