Learn More
Isofield™ Peta 230 Non-Sterile Earloop Cleanroom Facemasks with Clip - ISO Class 4/5 Compatible
BIOPHARMA
Product Code. 30178900
Change view
Click to view available options
Size:
Wide
Unit Size:
Pack of 600
Product Code. Size Unit Size
30178900 Wide Pack of 600
1 options
Product Code. 30178900 Supplier Isofield™ Supplier No. 60210
Estimated Shipment: 18-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Isofield Peta is ISO Class 4 compatible. The Peta is also fitted with a convenient clip to adapt to your SOP. The individually packed Peta has excellent bacterial and particle filtration efficiency, and an excellent choice for GMP Grade C/D environments.
  • Non-sterile earloop cleanroom facemask with clip
  • 230 mm length with malleable nose strip
  • 3-layer non-woven configuration
  • Highly efficient bacterial and particle filtration
  • Particle Shedding (Helmke Drum) Category I
  • Individually packed to maintain low CFU count
  • Pouches printed with IPA resistant ink to maintain traceability
Specifications
Product Type Cleanroom Face Mask
Color White
Fastening Type Earloops
Material Polyethylene, Polypropylene
Size Wide
Sterility Non-sterile
Style Pleated