missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEX5 Antibody (OTI6E9), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44637
This item is not returnable.
View return policy
Description
PEX5 Monoclonal antibody specifically detects PEX5 in Canine,Human,Monkey,Mouse,Rat samples. It is validated for Flow Cytometry,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| PEX5 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized powder ( 1X PBS, pH 7.3,) | |
| Mouse | |
| >95% | |
| RUO | |
| 5830 | |
| Canine, Human, Monkey, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| OTI6E9 | |
| Western Blot 1:500-2000, Flow Cytometry 1:100, Immunocytochemistry/ Immunofluorescence 1:100 | |
| FLJ50634, FLJ50721, Peroxin-5, peroxisomal biogenesis factor 5, Peroxisomal C-terminal targeting signal import receptor, peroxisomal targeting signal 1 receptor, peroxisomal targeting signal import receptor, peroxisomal targeting signal receptor 1, Peroxisome receptor 1peroxin-5, PTS1 receptor, PTS1-BP, PTS1RFLJ51948, PXR1peroxisomal targeting signal 1 (SKL type) receptor | |
| Full length human recombinant protein of human PEX5 (NP_000310) produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| For reconstitution, we recommend adding 100uL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiment, we strongly recommend performing another round of desalting process. | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction