missing translation for 'onlineSavingsMsg'
Learn More

PF-04691502, MedChemExpress

Product Code. 16482457
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482457 50 mg 50mg
16456656 100 mg 100mg
16446348 10 mg 10mg
16418937 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16482457 Supplier Medchem Express Supplier No. HY15177
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress PF-04691502 is a potent and selective inhibitor of PI3K and mTOR. PF-04691502 binds to human PI3Kα, β, δ, γ and mTOR with Kis of 1.8, 2.1, 1.6, 1.9 and 16 nM, respectively.

PF-04691502 is a potent and selective inhibitor of PI3K and mTOR. PF-04691502 binds to human PI3Kα, β, δ, γ and mTOR with Kis of 1.8, 2.1, 1.6, 1.9 and 16 nM, respectively.

Specifications

Chemical Name or Material PF-04691502
Physical Form Solid
CAS 1013101-36-4
Color Gray
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C22H27N5O4
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : 50 mg/mL (117.51 mM; Need ultrasonic)
SMILES OCCO[C@@H]1CC[C@@H](N2C(N=C(N)N=C3C)=C3C=C(C4=CC=C(OC)N=C4)C2=O)CC1
Molecular Weight (g/mol) 425.48
Formula Weight 425.48
Percent Purity 98.89%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.