missing translation for 'onlineSavingsMsg'
Learn More

PF-05175157, MedChemExpress

Product Code. 16485438
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16485438 10 mg 10mg
16481617 50 mg 50mg
16433346 25 mg 25mg
16428157 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16485438 Supplier Medchem Express Supplier No. HY12942
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress PF-05175157 is broad spectrum acetyl-CoA carboxylase (ACC) inhibitor with IC50s of 27.0, 33.0, 23.5 and 50.4 nM for ACC1 (human), ACC2 (human), ACC1 (rat), ACC2 (rat), respectively.

PF-05175157 is broad spectrum acetyl-CoA carboxylase (ACC) inhibitor with IC50s of 27.0, 33.0, 23.5 and 50.4 nM for ACC1 (human), ACC2 (human), ACC1 (rat), ACC2 (rat), respectively.

Specifications

Chemical Name or Material PF-05175157
Physical Form Solid
CAS 1301214-47-0
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C23H27N5O2
For Use With (Application) Metabolism-sugar/lipid metabolism
Solubility Information DMSO : 30 mg/mL (73.98 mM; Need ultrasonic and warming)
SMILES O=C1CC2(CCN(C(C3=CC=C4C(NC(C)=N4)=C3)=O)CC2)CC5=C1N(C(C)C)N=C5
Molecular Weight (g/mol) 405.49
Formula Weight 405.49
Percent Purity 96.13%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.