missing translation for 'onlineSavingsMsg'
Learn More

PF-573228, MedChemExpress

Product Code. 16463988
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16463988 10 mg 10mg
16414996 100 mg 100mg
16410347 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16463988 Supplier Medchem Express Supplier No. HY10461
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress PF-573228 is a potent and selective FAK inhibitor with IC50 of 4 nM for purified recombinant catalytic fragment of FAK.

PF-573228 is a potent and selective FAK inhibitor with IC50 of 4 nM for purified recombinant catalytic fragment of FAK.

Specifications

Chemical Name or Material PF-573228
CAS 869288-64-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H20F3N5O3S
Quantity 10 mg
Solubility Information DMSO : 25 mg/mL (50.87 mM; Need ultrasonic)
SMILES O=C1NC2=C(C=C(NC3=NC=C(C(F)(F)F)C(NCC4=CC=CC(S(=O)(C)=O)=C4)=N3)C=C2)CC1
Molecular Weight (g/mol) 491.49
Formula Weight 491.49
Percent Purity 98.09%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.