missing translation for 'onlineSavingsMsg'
Learn More

PFAA Mixture 100 μg/mL in Methanol:Water, Dr. Ehrenstorfer

Product Code. 30325006
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30325006 5 x 1 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30325006 Supplier Dr. Ehrenstorfer Supplier No. DREGS09000218MW
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. Se returpolicy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifikationer

CAS 375-73-5, 335-76-2, 307-55-1, 375-85-9, 307-24-4, 375-95-1
Quantity 5 x 1 mL
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol/Water
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.