missing translation for 'onlineSavingsMsg'
Learn More

PFK-158, MedChemExpress

Product Code. 16465018
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16465018

Brand: Medchem Express HY12203

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PFK-158 is a potent and selective PFKFB3 inhibitor with an IC50 value 137 nM. PFK-158 reduces glucose uptake, ATP production, lactate release, and induces apoptosis and autophagy in cancer cells. PFK-158 has broad anti-tumor activity. PFK-158 can also enhance Colistin's resistance to bacteria.

PFK-158 is a potent and selective PFKFB3 inhibitor with an IC50 value 137 nM. PFK-158 reduces glucose uptake, ATP production, lactate release, and induces apoptosis and autophagy in cancer cells. PFK-158 has broad anti-tumor activity. PFK-158 can also enhance Colistin's resistance to bacteria.

Specifications

Chemical Name or Material PFK-158
Physical Form Solid
CAS 1462249-75-7
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C18H11F3N2O
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 30 mg/mL (91.38 mM)
SMILES O=C(C1=CC=NC=C1)/C=C/C2=NC3=CC(C(F)(F)F)=CC=C3C=C2
Molecular Weight (g/mol) 328.29
Formula Weight 328.29
Percent Purity 98.52%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.