missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PFKFB2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 30425-1-AP-20UL
This item is not returnable.
View return policy
Description
The PFKFB2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PFKFB2. This antibody recognizes human antigen. The PFKFB2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| PFKFB2 | |
| 500 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| 6PF 2 K/Fru 2,6 P2ase 2, DKFZp781D2217, PFK 2/FBPase 2, PFK/FBPase 2, PFKFB2 | |
| 20 μL | |
| 5208 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction