missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PGA5 293T Cell Transient Overexpression Lysate
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.01mL
Specifications
Specifications
| Quantity | 0.1 mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| Research Category | Cancer, Cell Biology, Endocrinology, Signal Transduction |
| Target | PGA5 |
| Synonym | EC 3.4.23, EC 3.4.23.1, pepsin A, pepsinogen 5, group I (pepsinogen A), pepsinogen A5, PGA3, PGA4 |
| Gene Symbol | PGA5 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction