missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGAM5 Antibody (CL0624) [Alexa Fluor™ 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52947AF350
This item is not returnable.
View return policy
Description
PGAM5 Monoclonal antibody specifically detects PGAM5 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown
Specifications
| PGAM5 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| Bcl-XL-binding protein v68, BXLBv68, EC 3.1.3.16, MGC5352, phosphoglycerate mutase family member 5BXLBV68, serine/threonine-protein phosphatase PGAM5, mitochondrial | |
| This antibody was developed using a recombinant protein derived from Q96HS1, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Signal Transduction | |
| 192111 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown | |
| CL0624 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction