Learn More
PGC1a Mouse anti-Human, Unconjugated, Clone: 1C1B2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66369-1-IG-150UL
 View more versions of this product
Product Code. 16828185
  • 410.00 € / 150µL
Description
The PGC1a antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human PGC1a. This antibody recognizes human, mouse, rat antigen. The PGC1a antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Specifications
PGC1a
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence, ELISA
1C1B2
Unconjugated
150 μL
10891
IgG1