missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGF Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA519193
This item is not returnable.
View return policy
Description
This antibody is predicted to react with porcine based on sequence homology.
PGF Polyclonal antibody specifically detects PGF in Human samples. It is validated for Western Blot.
Specifications
| PGF | |
| Polyclonal | |
| Unconjugated | |
| PGF | |
| AI854365; D12S1900; PGF; PGFL; PIGF; Placenta growth factor; placental growth factor; placental growth factor, vascular endothelial growth factor-related protein; Plgf; PlGF 1; PlGF-2; SHGC; SHGC-10760 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 5228 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P49763 | |
| PGF | |
| Synthetic peptide sequence (NVTMQLLKIRSGDR) corresponding to the internal amino acids of PGF | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction