missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGII Monoclonal Antibody (OTI5C9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809596
This item is not returnable.
View return policy
Description
Hydrolyzes a variety of proteins.Specifications
| PGII | |
| OTI5C9 | |
| Unconjugated | |
| PGC | |
| PEPC, PGII | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5225 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P20142 | |
| PGC | |
| Human recombit protein fragment corresponding to amino acids 17-388 of human PGII produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction