missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGII Monoclonal Antibody (OTI5C9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809596S
This item is not returnable.
View return policy
Description
PGII Monoclonal antibody specifically detects PGII in Human samples. It is validated for LuminexSpecifications
| PGII | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P20142 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 5225 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Luminex | |
| OTI5C9 | |
| Unconjugated | |
| Pgc | |
| 2210410L06Rik; Gastricsin; pep carboxylase; PEPC; pepcase; pepsin C; pepsinogen C; pepsinogen group II; PG1; Pg-1; Pgc; PGII; preprogastricsin; progastricsin; progastricsin (pepsinogen C); Upg1; Upg-1; Urinary pepsinogen 1 | |
| Human recombinant protein fragment corresponding to amino acids 17-388 of human PGII produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction