missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGLYRP2/PGRP-L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
208.00 € - 382.00 €
Specifications
| Antigen | PGLYRP2/PGRP-L |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 2 μg/mL, Immunocytochemistry/ Immunofluorescence 10 - 20 μg/mL |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18698606
|
Novus Biologicals
NBP2-49690SS |
0.025 mg |
208.00 €
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18648987
|
Novus Biologicals
NBP2-49690 |
0.1 mg |
382.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PGLYRP2/PGRP-L Polyclonal antibody specifically detects PGLYRP2/PGRP-L in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| PGLYRP2/PGRP-L | |
| Western Blot 2 μg/mL, Immunocytochemistry/ Immunofluorescence 10 - 20 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| Q96PD5, Q96PD5, Q96PD5 | |
| 114770 | |
| IgG | |
| Protein G purified |
| 1.0 mg/mL | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Immunology, Inflammation | |
| PBS | |
| EC 3.5.1.28, peptidoglycan recognition protein 2tagl-beta, peptidoglycan recognition protein L, PGLYRPLHMFT0141, PGRP-LN-acetylmuramoyl-L-alanine amidase, PGRPLPeptidoglycan recognition protein long, tagL, tagL-alpha, TAGL-like | |
| Two synthetic peptides made to internal portions of the human PGLYRP2 protein (between amino acids 400-450 and 500-576) [UniProt Q96PD5] | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title