missing translation for 'onlineSavingsMsg'
Learn More

pH 12.00 Buffer Solution at 25°C, Reagecon
BIOPHARMA

Product Code. 11424569 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11424569 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11424569 Supplier Reagecon Supplier No. 1120525

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Tested, certified, NIST traceable, and shelf-stable buffer solution.

  • pH Buffer Reference Standard, pH Buffer Reference Material
  • Tested and certified by an ISO17025 Accredited Test Method (INAB Ref: 264T) and NIST traceable.
  • Guaranteed stability throughout its entire shelf life, even after opening the bottle.

Specifications

Description Accuracy ± 0.05 pH units
pH 12.00 ±0.05
Physical Form Liquid
Quantity 500 mL
Packaging HDPE Bottle GL45
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.