missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- pH Buffer Reference Standard, pH Buffer Reference Material
- Tested and certified by an ISO17025 Accredited Test Method (INAB Ref: 264T) and NIST traceable.
- Guaranteed stability throughout its entire shelf life, even after opening the bottle.
Specifications
Specifications
| Color | Clear |
| Description | Accuracy ± 0.01 pH units |
| pH | 4.00 ±0.01 |
| Physical Form | Liquid |
| Quantity | 1 L |
| Packaging | HDPE Bottle GL45 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
