missing translation for 'onlineSavingsMsg'
Learn More

pH 4 Buffer Standard, SPEX CertiPrep™

Product Code. 16391206 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16391206 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16391206 Supplier SPEX Certiprep Supplier No. PHBUFF4500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
pH4 SI Units

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material pH 4 Buffer Standard
Concentration 4 mol/L
For Use With (Application) pH Calibration
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Physical Form Liquid
Quantity 500 mL
Solution Type Standard
Packaging Triple-leached LDPE bottle
Concentration or Composition (by Analyte or Components) Various

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.