missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH 10.01 Calibration Buffer Sachets, Hanna Instruments™
Description
- Premium quality pH 10.01 calibration buffer that is NIST traceable, supplied with a Certificate of Analysis, and produced in accordance with ISO 3696/BS3978 standards
- Uses high purity salts, deionized water, certified weight-checked balances, and Class A glassware in a temperature controlled environment monitored with certified thermometers
- Reported values are accurate to +/- 0.01 pH @ 25°C and are traceable to NIST Standard Reference Materials (SRMs)
Specifications
Specifications
| Color | Undesignated |
| pH | 10.01 |
| Physical Form | Liquid |
| Quantity | 20 mL |
| Formulation | Aqueous Buffer Solution |
| Odor | Odorless |
| Packaging | 25Pack |
| Traceability to NIST | Yes |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?