missing translation for 'onlineSavingsMsg'
Learn More

pH 7.01 Calibration Buffer Sachets, Hanna Instruments™

Product Code. 10779061
Change view
Click to view available options
Quantity:
20 mL
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10779061 20 mL Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10779061 Supplier Hanna Instruments Supplier No. HI70007C
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Premium quality pH 7.01 calibration buffer that is NIST traceable and supplied with a Certificate of Analysis with an accuracy of +/- 0.01 pH @ 25°C.

  • Premium quality pH 7.01 calibration buffer that is NIST traceable and supplied with a Certificate of Analysis
  • Accuracy of +/- 0.01 pH @ 25°C
  • Temperature chart of the actual pH value at various temperatures printed on each sachet
  • pH 7.01 calibration buffer produced in accordance with ISO 3696/BS3978 standards
  • Uses high purity salts, deionized water, certified weight-checked balances, and Class A glassware 
  • In a temperature controlled environment monitored with certified thermometers
  • Reported values are accurate to +/- 0.01 pH @ 25°C and are traceable to NIST Standard Reference Materials (SRMs)
     

Specifications

Color Undesignated
pH 7.01
Physical Form Liquid
Quantity 20 mL
Formulation Aqueous Buffer Solution
Odor Odorless
Packaging 25Pack
Traceability to NIST Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.