missing translation for 'onlineSavingsMsg'
Learn More

Technical Buffer Solution pH 4, 7, and 10, SI Analytics™

Product Code. 11984769 Shop All SI Analytics Products
Click to view available options
Quantity:
60 Ampules
Unit Size:
Pack of 60
This item is not returnable. View return policy

Product Code. 11984769

Brand: SI Analytics 285138895

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

SI Analytics™ Technical Buffer Solution pH 4, 7, and 10 resists changes in pH when small quantities of an acid or an alkali are added to it.

  • Includes manufacturer certificate
  • Mixture, consisting of the following components: Water, sodium carbonate, sodium hydrogencarbonate
  • The product does not contain any relevant quantities of materials with critical values that have to be monitored at the workplace
  • Absorb with liquid-binding material (sand, diatomite, acid binders, universal binders, sawdust)
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 100°C
Color Amber
Density 1.01g/cm3
pH 4, 7, 10
Physical Form Liquid
Quantity 60 Ampules
Formulation Water, sodium carbonate, sodium hydrogencarbonate
Odor Odorless
Packaging FIOLAX™ Ampules
Sterility Sterile
Vapor Pressure 23hPa
Solubility Information Fully miscible in water
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.