missing translation for 'onlineSavingsMsg'
Learn More

pH Buffer Sachets, METTLER TOLEDO™

Product Code. 10431721
Change view
Click to view available options
Quantity:
30 x 20 mL
pH:
10.01
4, 7, 10
Unit Size:
597mL
600mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity pH Unit Size
10431721 30 x 20 mL 10.01 600mL
12683516 30 x 20 mL 10.01 597mL
12693516 30 x 20 mL 4, 7, 10 600mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10431721 Supplier Mettler Toledo Online Supplier No. 51302079
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Buffer solutions in sealed sachets for easy and precise calibration of pH measurement equipment.

  • Traceable to NIST primary standards
TRUSTED_SUSTAINABILITY

Specifications

Color Blue/White
Concentration 0.02 M
For Use With (Application) Calibration
For Use With (Equipment) pH Meter
pH 10.01
Physical Form Liquid
Quantity 30 x 20 mL
Solution Type Buffer
Packaging Satchets
Grade Laboratory
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.