missing translation for 'onlineSavingsMsg'
Learn More

pH Buffer Sachets, METTLER TOLEDO™
GREENER_CHOICE

Product Code. 12693516
Change view
Click to view available options
Quantity:
30 x 20 mL
pH:
10.01
4, 7, 10
Unit Size:
597mL
600mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity pH Unit Size
12693516 30 x 20 mL 4, 7, 10 600mL
10431721 30 x 20 mL 10.01 600mL
12683516 30 x 20 mL 10.01 597mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12693516 Supplier Mettler Toledo Online Supplier No. 51302080
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Buffer solutions in sealed sachets for easy and precise calibration of pH measurement equipment.

  • Traceable to NIST primary standards
TRUSTED_SUSTAINABILITY

Specifications

Color Multicolored
For Use With (Application) Calibration
For Use With (Equipment) pH Meter
pH 4, 7, 10
Physical Form Powder or Solid
Quantity 30 x 20 mL
Solution Type pH Buffer
Packaging Satchets
Grade Technical
compliance-icons
Product Identifier
  • Mettler-Toledo Rainbow II buffer, pH 4.01/7.00/10.00, 3 packs of 10x20 mL sachets
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: sodium hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.