missing translation for 'onlineSavingsMsg'
Learn More

Jenway™ pH Buffer Solution

Product Code. 11460479
Click to view available options
Quantity:
10 x 12 mL
pH:
7
10
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 11460479

Brand: Jenway 1615689

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Offers a range of pH buffers to care for your Jenway pH electrodes. Jenway™ ph Buffer Solution also offers a range of storage solutions and fill solutions tailored to your specific probe.

Offers a range of pH buffers to care for your Jenway pH electrodes. There is also a range of storage solutions and fill solutions tailored to your specific probe.

Specifications

For Use With (Equipment) pH Electrode
pH 7
Physical Form Liquid
Quantity 10 x 12 mL
Packaging 12mL Sachet
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.