missing translation for 'onlineSavingsMsg'
Learn More

Jenway™ pH Buffer Solution

Product Code. 11470479
Change view
Click to view available options
Quantity:
10 x 12 mL
pH:
7
10
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity pH unitSize
11470479 10 x 12 mL 10 Pack of 10
11460479 10 x 12 mL 7 Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11470479 Supplier Jenway Supplier No. 1615690

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Offers a range of pH buffers to care for your Jenway pH electrodes. Jenway™ ph Buffer Solution also offers a range of storage solutions and fill solutions tailored to your specific probe.

Offers a range of pH buffers to care for your Jenway pH electrodes. There is also a range of storage solutions and fill solutions tailored to your specific probe.

Specifications

For Use With (Equipment) pH Electrode
pH 10
Physical Form Liquid
Quantity 10 x 12 mL
Packaging 12mL Sachet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.