missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PHA 665752

Product Code. 18025072
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18025072

Brand: Tocris Bioscience™ 2693/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective MET inhibitor

Potent, selective and ATP-competitive inhibitor of MET kinase (IC50 values are 9, 68, 200, 1400, 3000, 3800 and 6000 nM for MET, Ron, Flk-1, c-abl, FGFR1, EGFR and c-src respectively and > 10000 nM for IGF-IR, PDGFR, AURORA2, PKA, PKBα, p38α, MK2 and MK3). Antitumor agent; inhibits tumorigenicity and angiogenesis in mouse lung cancer xenografts.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 477575-56-7
Molecular Formula C32H34Cl2N4O4S

Specifications

Quantity 10 mg
Formula Weight 641.61
Purity >98%
Molecular Weight (g/mol) 641.608
Chemical Name or Material PHA 665752
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.