missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phafin 1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555268
This item is not returnable.
View return policy
Description
Immunogen sequence: MVDHLANTEI NSQRIAAVES CFGASGQPLA LPGRVLLGEG VLTKECRKKA KPRIFFLFND ILVYGSIVLN KRKYRSQHII PLEEVTLELL PETLQAKNRW MIKTAKKSFV VSAASATERQ EWISHIEE Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 97%.
APPD (Apoptosis-inducing protein D; phafin 1) is predicted through gene annotation and contains pleckstrin homology domain. It may binds to two Zn++ ions through its FYVE zinc finger domain. Biological function of this protein has not been clearly mapped yet.
Specifications
| Phafin 1 | |
| Polyclonal | |
| Unconjugated | |
| PLEKHF1 | |
| 1810013P09Rik; AI037048; Apoptosis-inducing protein; apoptosis-inducing protein D; APPD; Lapf; lysosome-associated apoptosis-inducing protein containing PH and FYVE domains; PH and FYVE domain-containing protein 1; PH domain-containing family F member 1; phafin 1; PHAFIN1; phafin-1; pleckstrin homology and FYVE domain containing 1; pleckstrin homology domain containing, family F (with FYVE domain) member 1; pleckstrin homology domain-containing family F member 1; Plekhf1; SGP-1; ZFYVE15; Zinc finger FYVE domain-containing protein 15 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79156 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96S99 | |
| PLEKHF1 | |
| Recombinant protein corresponding to Human Phafin 1. Recombinant protein control fragment (Product #RP-89557). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction