missing translation for 'onlineSavingsMsg'
Learn More

Phenacetin, MedChemExpress

Product Code. 16470948
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16470948 5 g 5g
16402199 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16470948 Supplier Medchem Express Supplier No. HYB0476
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Phenacetin (Acetophenetidin) is a non-opioid analgesic/antipyretic agent. Phenacetin is a selective COX-3 inhibitor. Phenacetin is used as probe of cytochrome P450 enzymes CYP1A2 in human liver microsomes and in rats.

Phenacetin (Acetophenetidin) is a non-opioid analgesic/antipyretic agent. Phenacetin is a selective COX-3 inhibitor. Phenacetin is used as probe of cytochrome P450 enzymes CYP1A2 in human liver microsomes and in rats.

Specifications

Chemical Name or Material Phenacetin
CAS 62-44-2
Color White
Purity Grade Notes Research
Molecular Formula C10H13NO2
Quantity 5 g
Synonym Acetophenetidin
Solubility Information DMSO : ≥ 100 mg/mL (557.97 mM) ∣H2O : 0.67 mg/mL (3.74 mM; Need ultrasonic)
SMILES CC(NC1=CC=C(OCC)C=C1)=O
Molecular Weight (g/mol) 179.22
Formula Weight 179.22
Percent Purity 99.54%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.