missing translation for 'onlineSavingsMsg'
Learn More

Phenacetin, TRC

Product Code. 30133251
Change view
Click to view available options
Quantity:
50 mg
1 g
5 g
Unit Size:
1g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133251 1 g 1g
18093240 50 mg 50mg
18092120 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133251 Supplier Toronto Research Chemicals Supplier No. TRCP2945801G
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-44-2
Molecular Formula C10 H13 N O2
Molecular Weight (g/mol) 179.22
Synonym Phenacetin, Acetamide, N-(4-ethoxyphenyl)-, p-Acetophenetidide (8CI), N-(4-Ethoxyphenyl)acetamide, 4-(Acetylamino)phenetole, 4-Ethoxy-1-acetylaminobenzene, 4-Ethoxyacetanilide, 4'-Ethoxyacetanilide, Aceto-4-phenetidine, Acetophenetidin, Acetophenetidine, Acetophenetin, Acetparaphenetidine, Acetphenetidin, Fenidina, Fenina, Kalmin, N-Acetyl-4-ethoxyaniline, N-Acetyl-p-ethoxyaniline, N-Acetyl-p-phenetidine, NSC 7651, Pertonal, Phenacetine, Phenazetin, Phenedina, Phenidin, Phenin, p-Ethoxyacetanilide, p-Phenetidin acetate
IUPAC Name N-(4-ethoxyphenyl)acetamide
SMILES CCOc1ccc(NC(=O)C)cc1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H13NO2/c1-3-13-10-6-4-9(5-7-10)11-8(2)12/h4-7H,3H2,1-2H3,(H,11,12)
Formula Weight 179.0946
Percent Purity >95%
Chemical Name or Material Phenacetin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.