missing translation for 'onlineSavingsMsg'
Learn More

Phenanthrene-9-carboxaldehyde, 97%

Product Code. 11491667
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491667 1 g 1g
11401677 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11491667 Supplier Thermo Scientific Alfa Aesar Supplier No. B25648.03
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 4707-71-5 | C15H10O | 206.24 g/mol

Phenanthrene-9-carboxaldehyde is used to make fluorescent probes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenanthrene-9-carboxaldehyde is used to make fluorescent probes.

Solubility
Soluble in acetone, dichloromethane, methanol.

Notes
Air sensitive. Store away from oxidizing agents, air. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4707-71-5
Molecular Formula C15H10O
Molecular Weight (g/mol) 206.24
MDL Number MFCD00001175
InChI Key QECIGCMPORCORE-UHFFFAOYSA-N
Synonym 9-phenanthrenecarboxaldehyde, phenanthrene-9-carboxaldehyde, 9-formylphenanthrene, phenanthrene-9-aldehyde, 9-phenanthrenecarbaldehyde, 9-phenanthrenealdehyde, 9-phenanthrencarbaldehyd, 9-phenanthraldehyde, 9-phenanthrenaldehyde, phenanthren-9-aldehyd
PubChem CID 78437
IUPAC Name phenanthrene-9-carbaldehyde
SMILES O=CC1=CC2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 100°C to 103°C
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Soluble in acetone,dichloromethane,methanol.
Formula Weight 206.24
Percent Purity 97%
Chemical Name or Material Phenanthrene-9-carboxaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.