missing translation for 'onlineSavingsMsg'
Learn More

Phenol, 99+%, ACS reagent, ACROS Organics™

Product Code. 10095640 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
25g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10095640 25g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10095640 Supplier Thermo Scientific Acros Supplier No. 417170250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 108-95-2
Melting Point >40.5°C (on dry substance)
Color Colorless to White
Density 1.0700g/mL
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass Bottle
Molecular Formula C6H6O
Linear Formula C6H5OH
MDL Number MFCD00002143
Quantity 25 g
Beilstein 06, 110
Residue after Evaporation 0.05% max.
Fieser 01,828; 13,166; 14,88
Merck Index 15, 7354
Synonym carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
SMILES OC1=CC=CC=C1
IUPAC Name phenol
Molecular Weight (g/mol) 94.11
PubChem CID 996
ChEBI CHEBI:15882
Formula Weight 94.11
Percent Purity ≥99.0%
Grade ACS Reagent
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Phenol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.