missing translation for 'onlineSavingsMsg'
Learn More

Phenol, 99+%, for biochemistry, loose crystals

Product Code. 10113820 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10113820 500 g Glass bottle 500g
10615442 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10113820 Supplier Thermo Scientific Acros Supplier No. 180785000
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 108-95-2 | C6H6O | 94.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-95-2
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.11
MDL Number MFCD00002143
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
Synonym carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
PubChem CID 996
ChEBI CHEBI:15882
IUPAC Name phenol
SMILES OC1=CC=CC=C1

Specifications

Melting Point 39.0°C to 42.0°C
Color Colorless to White
Density 1.0700g/mL
Boiling Point 182.0°C
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C6H5OH
Quantity 500 g
Beilstein 06, 110
Residue after Evaporation 0.05% max.
Fieser 01,828; 13,166; 14,88
Merck Index 15, 7354
Specific Gravity 1.07
Solubility Information Solubility in water: 8.4g/L (20°C). Other solubilities: soluble in aq. alkali hydroxides, soluble in alcohol, chloroform, ether, glycerol,, carbon disulfide and fixed oils, 1g/12mL benzene, practically insoluble in petroleum ether
Viscosity 3.437 mPa.s (50°C)
Formula Weight 94.11
Percent Purity 99+%
Grade Biochemistry
Physical Form Loose Crystals
Chemical Name or Material Phenol, 99+%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H341-Suspected of causing genetic defects.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.