missing translation for 'onlineSavingsMsg'
Learn More

Phenol/Chloroform/Isoamyl Alcohol (125:24:1 Mixture, pH 4.3, Liq.), Fisher BioReagents™

Product Code. 10205113
Change view
Click to view available options
Quantity:
100 mL
400 mL
Packaging:
Coated Amber Glass/Poison Pack
Unit Size:
100mL
400mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10205113 100 mL Coated Amber Glass/Poison Pack 100mL
10699543 400 mL Coated Amber Glass/Poison Pack 400mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10205113 Supplier Fisher BioReagents Supplier No. BP1754I100
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Phenol/Chloroform/Isoamyl Alcohol (125:24:1 Mixture, pH 4.3, Liq.)

This mixture is used to purify total RNA from mixtures containing RNA, DNA, and protein.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-51-3
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
InChI Key PHTQWCKDNZKARW-UHFFFAOYSA-N
Synonym 3-methyl-1-butanol, isoamyl alcohol, isopentyl alcohol, isopentanol, 3-methylbutanol, 1-butanol, 3-methyl, isoamylol, isobutylcarbinol, iso-amylalkohol, isobutyl carbinol
PubChem CID 31260
ChEBI CHEBI:15837
IUPAC Name 3-methylbutan-1-ol
SMILES CC(C)CCO

Specifications

Merck Index 15, 7354
Percent Purity ≥99%
Identification Pass Test
Packaging Coated Amber Glass/Poison Pack
pH 4.3
Quantity 100 mL
Physical Form Liquid
Chemical Name or Material Phenol/Chloroform/Isoamyl Alcohol, 125:24:1 Mixture
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.