missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenol/Chloroform/Isoamyl Alcohol (25:24:1 Mixture, pH 5.2±0.3, Liq.), Fisher BioReagents
Description
This mixture is used for extracting protein from nucleic acid preparations.
Specifications
Specifications
| Chemical Name or Material | Phenol/Chloroform/Isoamyl Alcohol |
| Boiling Point | 96°C |
| Molecular Formula | CHCl3 |
| MDL Number | MFCD00000826 |
| Merck Index | 15, 7354 |
| Synonym | trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio |
| InChI Key | HEDRZPFGACZZDS-UHFFFAOYSA-N |
| SMILES | ClC(Cl)Cl |
| IUPAC Name | trichloromethane |
| Molecular Weight (g/mol) | 119.37 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?