missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Phenol:Chloroform:Isoamyl alcohol 25:24:1, Ready-to-Use saturated aq. soln., pH 6.7, with alkaline buffer

Product Code. 15465785
Change view
Click to view available options
Quantity:
100 mL
400 mL
Unit Size:
100mL
400mL
Product Code. Quantity unitSize
15465785 400 mL 400mL
15455785 100 mL 100mL
2 options
This item is not returnable. View return policy

Product Code. 15465785

Brand: Thermo Scientific Chemicals J60331.AN

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Useful for purifying RNA | CAS: 136112-00-0 | C12H19Cl3O2 | 301.632 g/mol

Phenol:Chloroform:Isoamyl alcohol 25:24:1 is used in the extraction of DNA for the generation of DNA minisatellite fingerprints. It is also used for purifying RNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Note: Buffer is sold separately; see 327345000.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136112-00-0
Molecular Formula C12H19Cl3O2
Molecular Weight (g/mol) 301.632
MDL Number MFCD00133763
InChI Key ZYWFEOZQIUMEGL-UHFFFAOYSA-N
Synonym phenol-chloroform-isoamyl alcohol mixture, chloroform; isoamyl alcohol; phenol, phenol chloroform isoamyl alcohol, phenol-chloroform-isoamyl alcohol mixture, bioultra, for molecular biology, 125:24:1, phenol-chloroform-isoamyl alcohol mixture, bioultra, for molecular biology, 25:24:1, phenol-chloroform-isoamyl alcohol mixture, vetec tm reagent grade, 25:24:1, phenol-chloroform-isoamyl alcohol mixture, bioultra, for molecular biology, 49.5:49.5:1, phenol:chloroform:isoamyl alcohol 25:24:1, ready-to-use saturated aqueous solution, ph 5.2, with alkaline buffer, phenol:chloroform:isoamyl alcohol 25:24:1, ready-to-use saturated aqueous solution, ph 6.7, with alkaline buffer
PubChem CID 66587205
IUPAC Name chloroform;3-methylbutan-1-ol;phenol
SMILES CC(C)CCO.C1=CC=C(C=C1)O.C(Cl)(Cl)Cl

Specifications

Color Colorless
UN Number UN2922
pH 6.7
Quantity 400 mL
Physical Form Liquid
Chemical Name or Material Phenol:Chloroform:Isoamyl alcohol 25:24:1, Ready-to-Use saturated aq. soln.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.