missing translation for 'onlineSavingsMsg'
Learn More

Phenol, detached crystals, 99+%

Product Code. 11432418
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11432418 500 g 500g
11422418 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11432418 Supplier Thermo Scientific Alfa Aesar Supplier No. A15760.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-95-2 | C6H6O | 94.11 g/mol

Phenol is used as a precursor for cyclohexanone, plastics, nonionic detergents and pharmaceutical drugs like aspirin. It acts as an anesthetic in chloraseptic. It reacts with acetone to get bisphenol-A, which is used in the synthesis of poly carbonates and epoxide resins. It is also used in the manufacture of synthetic resins, dyes, pharmaceuticals, synthetic tanning agents, perfumes, lubricating oils and solvents. In molecular biology, it is used in the extraction of nucleic acid from tissues by using liquid/liquid phenol-chloroform extraction technique. It is an active component of paint strippers, which is used for the removal of epoxy and polyurethane. It is also used in the preparation of cosmetics, hair colorings and skin lightening preparations. In the field of medicine, its spray is useful in helping sore throat.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water, alcohol, chloroform, ether, benzene, glycerol, acetone, carbon disulfide and aqueous alkali hydroxides.

Notes
Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong bases and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-95-2
Molecular Formula C6H6O
Molecular Weight (g/mol) 94.11
MDL Number MFCD00002143
InChI Key ISWSIDIOOBJBQZ-UHFFFAOYSA-N
Synonym carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol
PubChem CID 996
ChEBI CHEBI:15882
IUPAC Name phenol
SMILES OC1=CC=CC=C1

Specifications

Melting Point 40°C to 43°C
Density 1.071
Boiling Point 181°C
Flash Point 79°C (175°F)
Odor Sweet
Quantity 500 g
UN Number UN1671
Beilstein 969616
Sensitivity Air and light sensitive
Merck Index 14,7241
Solubility Information Soluble in water,alcohol,chloroform,ether,benzene,glycerol,acetone,carbon disulfide and aqueous alkali hydroxides.
Formula Weight 94.11
Percent Purity ≥99%
Chemical Name or Material Phenol, detached crystals
Afficher plus Afficher moins
compliance-icons compliance-icons compliance-icons
Identifiant produit
  • Phenol
Mention d'avertissement
  • Danger
Catégorie de danger
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
Mention de danger
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H341-Suspected of causing genetic defects.
  • H373-May cause damage to organs.
Conseils de prudence
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Informations supplémentaires
  • MIXTURE LIST-Contains : phenol

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.